COM448612982 Combi-Blocks, Inc.

(6-Butoxypyridin-2-yl)boronic acid

  • MDL number MFCD27935498
  • Cas Number : 2377607-55-9
  • Molecular Weight : 195.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0037

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0037
Availability details for
Quantity
  • Availability
  • Add to Cart