COM448612987 Combi-Blocks, Inc.

[(2,6-Dimethylphenyl)methyl]boronic acid

  • MDL number MFCD24265286
  • Cas Number : 2246670-18-6
  • Molecular Weight : 164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0048
Availability details for
Quantity
  • Availability
  • Add to Cart