COM448612988 Combi-Blocks, Inc.

(1-tert-Butylpyrazol-3-yl)boronic acid

  • MDL number MFCD27935502
  • Molecular Weight : 168.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0052

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0052
Availability details for
Quantity
  • Availability
  • Add to Cart