COM448612999 Combi-Blocks, Inc.

2-Chloro-4-fluoro-3-methylpheny)boronic acid

  • MDL number MFCD23712936
  • Cas Number : 2304633-76-7
  • Molecular Weight : 188.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0072

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0072
Availability details for
Quantity
  • Availability
  • Add to Cart