COM448613007 Combi-Blocks, Inc.

{6-Methylimidazo[1,2-a]pyridin-8-yl}boronic acid

  • MDL number MFCD27935517
  • Cas Number : 2377607-68-4
  • Molecular Weight : 176.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0082

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0082
Availability details for
Quantity
  • Availability
  • Add to Cart