COM448613009 Combi-Blocks, Inc.

[(3,4-Difluorophenyl)methyl]boronic acid

  • MDL number MFCD24332193
  • Cas Number : 2246751-44-8
  • Molecular Weight : 171.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0084

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0084
Availability details for
Quantity
  • Availability
  • Add to Cart