COM448613010 Combi-Blocks, Inc.

(2,4-Difluorophenyl)methylboronic acid

  • MDL number MFCD18861690
  • Cas Number : 1350513-42-6
  • Molecular Weight : 171.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0085

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0085
Availability details for
Quantity
  • Availability
  • Add to Cart