COM448613013 Combi-Blocks, Inc.

2-(1-Cyanoethyl)phenylboronic acid

  • MDL number MFCD27935520
  • Cas Number : 2377587-40-9
  • Molecular Weight : 175.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0088

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0088
Availability details for
Quantity
  • Availability
  • Add to Cart