COM448613017 Combi-Blocks, Inc.

{2-[(2-Methylpropyl)sulfanyl]pyrimidin-5-yl}boronic acid

  • MDL number MFCD27935524
  • Molecular Weight : 212.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0092

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0092
Availability details for
Quantity
  • Availability
  • Add to Cart