COM448613018 Combi-Blocks, Inc.

5-Methyl-[1,2,3,4]tetrazolo[1,5-a]pyridino-6-boronic acid

  • MDL number MFCD27935525
  • Cas Number : 2377607-57-1
  • Molecular Weight : 178.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0093

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0093
Availability details for
Quantity
  • Availability
  • Add to Cart