COM448613020 Combi-Blocks, Inc.

(2,5-Dichloro-6-methylpyridin-3-yl)boronic acid

  • MDL number MFCD27935527
  • Molecular Weight : 205.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0096
Availability details for
Quantity
  • Availability
  • Add to Cart