COM448613021 Combi-Blocks, Inc.

[(3-Fluoro-4-methylphenyl)methyl]boronic acid

  • MDL number MFCD27935528
  • Cas Number : 850040-35-6
  • Molecular Weight : 168.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0098
Availability details for
Quantity
  • Availability
  • Add to Cart