COM448613027 Combi-Blocks, Inc.

5-Fluoro-6-methylpyridine-2-boronic acid

  • MDL number MFCD08061580
  • Cas Number : 1208101-45-4
  • Molecular Weight : 154.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0109

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0109
Availability details for
Quantity
  • Availability
  • Add to Cart