COM448613028 Combi-Blocks, Inc.

[5-(Cyanomethyl)-2-fluorophenyl]boronic acid

  • MDL number MFCD27935530
  • Molecular Weight : 179.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0111
Availability details for
Quantity
  • Availability
  • Add to Cart