COM448613033 Combi-Blocks, Inc.

4-Chloropyridine-2-boronic acid

  • MDL number MFCD12401036
  • Cas Number : 870459-91-9
  • Molecular Weight : 157.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0153
Availability details for
Quantity
  • Availability
  • Add to Cart