COM448613037 Combi-Blocks, Inc.

{8-Methyl-[1,2,4]triazolo[4,3-a]pyridin-6-yl}boronic acid

  • MDL number MFCD27935535
  • Cas Number : 2377608-14-3
  • Molecular Weight : 177.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0301

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0301
Availability details for
Quantity
  • Availability
  • Add to Cart