COM448613041 Combi-Blocks, Inc.

{3-Ethyl-[1,2,3]triazolo[4,5-b]pyridin-6-yl}boronic acid

  • MDL number MFCD27935539
  • Molecular Weight : 192.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0307

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0307
Availability details for
Quantity
  • Availability
  • Add to Cart