COM448613046 Combi-Blocks, Inc.

[(2-Chloro-5-fluorophenyl)methyl]boronic acid

  • MDL number MFCD27935543
  • Cas Number : 2246614-18-4
  • Molecular Weight : 188.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0312
Availability details for
Quantity
  • Availability
  • Add to Cart