COM448613047 Combi-Blocks, Inc.

(3-Chloro-2,4-dimethoxyphenyl)boronic acid

  • MDL number MFCD23379571
  • Cas Number : 750585-61-6
  • Molecular Weight : 216.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0314

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0314
Availability details for
Quantity
  • Availability
  • Add to Cart