COM448613048 Combi-Blocks, Inc.

(3-Methyl-3,4-dihydro-2H-1,5-benzodioxepin-6-yl)boronic acid

  • MDL number MFCD27935544
  • Molecular Weight : 208.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0315

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0315
Availability details for
Quantity
  • Availability
  • Add to Cart