COM448613051 Combi-Blocks, Inc.

[3-(Methylsulfanyl)-[1,2,4]triazolo[4,3-a]pyridin-6-yl]boronic acid

  • MDL number MFCD27935547
  • Molecular Weight : 209.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0318

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0318
Availability details for
Quantity
  • Availability
  • Add to Cart