COM448613053 Combi-Blocks, Inc.

[(2-Fluoro-3-methoxyphenyl)methyl]boronic acid

  • MDL number MFCD27935549
  • Cas Number : 2246703-03-5
  • Molecular Weight : 184.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0320

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0320
Availability details for
Quantity
  • Availability
  • Add to Cart