COM448613054 Combi-Blocks, Inc.

{6-Methoxyimidazo[1,2-a]pyridin-2-yl}boronic acid

  • MDL number MFCD27935550
  • Molecular Weight : 192.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0321

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0321
Availability details for
Quantity
  • Availability
  • Add to Cart