COM448613056 Combi-Blocks, Inc.

[6-(2-Methoxyethoxy)-5-methylpyridin-3-yl]boronic acid

  • MDL number MFCD27935552
  • Cas Number : 2377608-87-0
  • Molecular Weight : 211.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0323

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0323
Availability details for
Quantity
  • Availability
  • Add to Cart