COM448613057 Combi-Blocks, Inc.

(5-Cyano-2,4-difluorophenyl)boronic acid

  • MDL number MFCD27935553
  • Molecular Weight : 182.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0324

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0324
Availability details for
Quantity
  • Availability
  • Add to Cart