COM448613059 Combi-Blocks, Inc.

{4-[(1E)-2-Cyanoeth-1-en-1-yl]-3-fluorophenyl}boronic acid

  • MDL number MFCD27935554
  • Molecular Weight : 191.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0326
Availability details for
Quantity
  • Availability
  • Add to Cart