COM448613066 Combi-Blocks, Inc.

[3-(Cyanomethoxy)-4-methoxyphenyl]boronic acid

  • MDL number MFCD27935561
  • Cas Number : 1033602-67-3
  • Molecular Weight : 207.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0333

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0333
Availability details for
Quantity
  • Availability
  • Add to Cart