COM448613073 Combi-Blocks, Inc.

(Naphthalen-1-ylmethyl)boronic acid

  • MDL number MFCD18861657
  • Cas Number : 86819-97-8
  • Molecular Weight : 186

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0340
Availability details for
Quantity
  • Availability
  • Add to Cart