COM448613079 Combi-Blocks, Inc.

(5-Chloro-4-fluoro-2-methoxyphenyl)boronic acid

  • MDL number MFCD23379570
  • Cas Number : 949892-09-5
  • Molecular Weight : 204.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0347

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0347
Availability details for
Quantity
  • Availability
  • Add to Cart