COM448613084 Combi-Blocks, Inc.

(2-Propylindazol-5-yl)boronic acid

  • MDL number MFCD27935574
  • Molecular Weight : 204.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0352

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0352
Availability details for
Quantity
  • Availability
  • Add to Cart