COM448613093 Combi-Blocks, Inc.

(7-Chloroquinoxalin-5-yl)boronic acid

  • MDL number MFCD27935583
  • Molecular Weight : 208.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0362
Availability details for
Quantity
  • Availability
  • Add to Cart