COM448613094 Combi-Blocks, Inc.

[5-Fluoro-2-(pyrrolidin-1-yl)phenyl]boronic acid

  • MDL number MFCD27935584
  • Cas Number : 2174971-35-6
  • Molecular Weight : 209.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0363

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0363
Availability details for
Quantity
  • Availability
  • Add to Cart