COM448613098 Combi-Blocks, Inc.

{6-Methylimidazo[1,2-a]pyridin-8-yl}boronic acid hydrochloride

  • MDL number MFCD27935588
  • Cas Number : 2377607-69-5
  • Molecular Weight : 212.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0367
Availability details for
Quantity
  • Availability
  • Add to Cart