COM448613102 Combi-Blocks, Inc.

[3-Methyl-5-(pyrrolidin-1-yl)phenyl]boronic acid

  • MDL number MFCD27935592
  • Molecular Weight : 205.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0371

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0371
Availability details for
Quantity
  • Availability
  • Add to Cart