COM448613111 Combi-Blocks, Inc.

[4-Fluoro-3-(piperidin-1-ylmethyl)phenyl]boronic acid

  • MDL number MFCD27935602
  • Cas Number : 1200131-35-6
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0384
Availability details for
Quantity
  • Availability
  • Add to Cart