COM448613115 Combi-Blocks, Inc.

{3,6-Dimethyl-[1,2,4]triazolo[4,3-a]pyridin-8-yl}boronic acid

  • MDL number MFCD27935608
  • Cas Number : 2377611-60-2
  • Molecular Weight : 191.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0390

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0390
Availability details for
Quantity
  • Availability
  • Add to Cart