COM448613116 Combi-Blocks, Inc.

(1-Ethyl-6-fluoro-1,2,3-benzotriazol-5-yl)boronic acid

  • MDL number MFCD27935609
  • Molecular Weight : 209.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0391
Availability details for
Quantity
  • Availability
  • Add to Cart