COM448613126 Combi-Blocks, Inc.

(1-Isopropylindazol-5-yl)boronic acid

  • MDL number MFCD27935618
  • Cas Number : 1551416-99-9
  • Molecular Weight : 204.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0404

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0404
Availability details for
Quantity
  • Availability
  • Add to Cart