COM448613129 Combi-Blocks, Inc.

(1-isopropylindol-5-yl)boronic acid

  • MDL number MFCD25957573
  • Cas Number : 1289141-89-4
  • Molecular Weight : 203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0407
Availability details for
Quantity
  • Availability
  • Add to Cart