COM448613131 Combi-Blocks, Inc.

[(4-tert-Butylphenyl)methyl]boronic acid

  • MDL number MFCD18861684
  • Cas Number : 1350513-29-9
  • Molecular Weight : 192.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0409

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0409
Availability details for
Quantity
  • Availability
  • Add to Cart