COM448613134 Combi-Blocks, Inc.

(2-Cyano-3-isopropoxyphenyl)boronic acid

  • MDL number MFCD27935624
  • Molecular Weight : 205.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Freezer

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0413
Availability details for
Quantity
  • Availability
  • Add to Cart