COM448613137 Combi-Blocks, Inc.

2-(Dihydroxyboranyl)-5-methoxypyridin-1-ium-1-olate

  • MDL number MFCD27935628
  • Molecular Weight : 168.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0417

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0417
Availability details for
Quantity
  • Availability
  • Add to Cart