COM448613138 Combi-Blocks, Inc.

2-(4-Methylpiperidin-1-yl)pyridine-5-boronic acid

  • MDL number MFCD22566420
  • Cas Number : 1864801-70-6
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0418

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0418
Availability details for
Quantity
  • Availability
  • Add to Cart