COM448613139 Combi-Blocks, Inc.

6-Cyano-5-(trifluoromethyl)pyridin-3-ylboronic acid

  • MDL number MFCD22491377
  • Cas Number : 2377606-68-1
  • Molecular Weight : 215.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0419

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0419
Availability details for
Quantity
  • Availability
  • Add to Cart