COM448613145 Combi-Blocks, Inc.

[3-(1,3-Oxazol-5-yl)phenyl]boronic acid

  • MDL number MFCD27935634
  • Cas Number : 902775-28-4
  • Molecular Weight : 189.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0427

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0427
Availability details for
Quantity
  • Availability
  • Add to Cart