COM448613146 Combi-Blocks, Inc.

[(5-Chloro-1-benzothiophen-3-yl)methyl]boronic acid

  • MDL number MFCD27935635
  • Molecular Weight : 226.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0428
Availability details for
Quantity
  • Availability
  • Add to Cart