COM448613148 Combi-Blocks, Inc.

(5,8-Difluoroquinolin-6-yl)boronic acid

  • MDL number MFCD27935636
  • Cas Number : 2260683-50-7
  • Molecular Weight : 209.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0430
Availability details for
Quantity
  • Availability
  • Add to Cart