COM448613149 Combi-Blocks, Inc.

(2,8-Dimethylquinolin-4-yl)boronic acid

  • MDL number MFCD27935637
  • Molecular Weight : 201.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0431
Availability details for
Quantity
  • Availability
  • Add to Cart