COM448613150 Combi-Blocks, Inc.

[6-Chloro-2-(methylsulfanyl)-1,3-benzothiazol-4-yl]boronic acid

  • MDL number MFCD27935638
  • Molecular Weight : 259.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0433

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0433
Availability details for
Quantity
  • Availability
  • Add to Cart