COM448613152 Combi-Blocks, Inc.

3-(Propylsulfonyl)phenylboronic acid

  • MDL number MFCD22199583
  • Cas Number : 1313617-52-5
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0435

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0435
Availability details for
Quantity
  • Availability
  • Add to Cart